Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data- Temperature and Humidity
For one free-selectable temperature and humidity value, and measuring of light intensity
31940.00 DKK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17119824
|
Memmert™
HPP410ecoD00137 |
31940.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
17129684
|
Memmert™
HPP110ecoD00137 |
HPP110eco |
31940.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17169774
|
Memmert™
HPP260ecoD00137 |
HPP260eco |
31940.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17199864
|
Memmert™
HPP750ecoD00137 |
HPP750eco |
31940.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Includes temperature distribution survey at Memmert
- For 27 measuring points to DIN 12880:2007-05
- PQ check list as support for validation by customer.
Specifications
IQ/OQ Document with Device-specific Works Test Data |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Memmert™ IQ/OQ Document with Device-Specific Works Test Data- Temperature and Humidity