Learn More
Memmert™ IQ/OQ Document with Device-Specific Works Test Data- Temperature and Humidity
For one free-selectable temperature and humidity value, and measuring of light intensity
31940.00 DKK
Specifications
Product Type IQ/OQ Document with Device-specific Works Test Data
View More Specs
Products 4
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
17119824
View Documents
Memmert™
HPP410ecoD00137
31940.00 DKK
Each
17129684
View Documents
Memmert™
HPP110ecoD00137
HPP110eco
31940.00 DKK
Each
17169774
View Documents
Memmert™
HPP260ecoD00137
HPP260eco
31940.00 DKK
Each
17199864
View Documents
Memmert™
HPP750ecoD00137
HPP750eco
31940.00 DKK
Each
Description
Description
  • Includes temperature distribution survey at Memmert
  • For 27 measuring points to DIN 12880:2007-05
  • PQ check list as support for validation by customer.
Specifications
Specifications
IQ/OQ Document with Device-specific Works Test Data
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ IQ/OQ Document with Device-Specific Works Test Data- Temperature and Humidity