missing translation for 'onlineSavingsMsg'
Learn More
Isopropyl methanesulfonate, 99% Product Code.: 10677191

Isopropyl methanesulfonate, 99%

Product Code. 10677191 Shop All Thermo Scientific Chemicals Products
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10677191

Brand: Thermo Scientific Acros 412840250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 926-06-7 | C4H10O3S | 138.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 926-06-7
Molecular Formula C4H10O3S
Molecular Weight (g/mol) 138.19
MDL Number MFCD00047802
InChI Key SWWHCQCMVCPLEQ-UHFFFAOYSA-N
Synonym isopropyl methanesulfonate, isopropyl mesylate, isopropylmethanesulfonate, methanesulfonic acid, 1-methylethyl ester, isopropyl methane sulphonate, 2-propyl methanesulphonate, isopropyl methane sulfonate, methanesulfonic acid, isopropyl ester, isopropyl methanesulphonate, unii-t0k2txy26bShow MoreShow Less
PubChem CID 13551
IUPAC Name propan-2-yl methanesulfonate
SMILES CC(C)OS(=O)(=O)C

Specifications

Color Colorless
Density 1.1450g/mL
Boiling Point 82.0°C (6.0 mmHg)
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3SO3CH(CH3)2
Refractive Index 1.4180 to 1.4210
Quantity 25 g
Beilstein 04,IV,13
Specific Gravity 1.145
Formula Weight 138.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Isopropyl methanesulfonate, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465676 Certificate TypeCertificate of Analysis Date23/08/2024 Catalog Number412840250,412840050,412841000
Lot NumberA0457407 Certificate TypeCertificate of Analysis Date03/11/2023 Catalog Number412840250,412840050,412841000
Lot NumberA0441702 Certificate TypeCertificate of Analysis Date07/06/2023 Catalog Number412840250,412840050,412841000
Lot NumberA0395808 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number412840250,412840050,412841000
Lot NumberA0409134 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number412840250,412840050,412841000
compliance-icons compliance-icons
Product Identifier
  • Isopropyl methanesulfonate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Isopropyl methanesulfonate, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.