missing translation for 'onlineSavingsMsg'
Learn More

GPE Scientific™ J Young Spherical Joint, Male/O-Ring Ball Joint

GPE Scientific™ J Young Spherical Joint, Male/O-Ring Ball Joint is an interchangeable spherical joint available with Viton or PTFE-covered Viton O-Ring.

Brand:  GPE Scientific™ GBM/18/7

 View more versions of this product

Product Code. 15804764

  • 180.00 DKK / Each
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Spherical joint designed for use with cup joint
  • Available with different joint sizes, shank bores, and shank ODs
Specifications

Specifications

Shank Bore: 7 mm, Shank OD: 11 mm
Male Spherical Joint
18/7
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.