Learn More
GPE Scientific™ J Young Spherical Joint, Male/O-Ring Ball Joint
GPE Scientific™ J Young Spherical Joint, Male/O-Ring Ball Joint is an interchangeable spherical joint available with Viton or PTFE-covered Viton O-Ring.
167.00 DKK - 489.00 DKK
Specifications
Type Male Spherical Joint
View More Specs
Products 9
Product Code Brand Type Joint Size Price Quantity & Availability  
Product Code Brand Type Joint Size Price Quantity & Availability  
15884754
View Documents
GPE Scientific™
GBM/12/2
Male Spherical Joint 12/2
252.00 DKK
Each
Estimated Shipment: 04-04-2025
to see stock.
Add to basket
15894754
View Documents
GPE Scientific™
GBM/12/5
Male Spherical Joint 12/5
167.00 DKK
Each
Estimated Shipment: 04-04-2025
to see stock.
Add to basket
15804764
View Documents
GPE Scientific™
GBM/18/7
Male Spherical Joint 18/7
180.00 DKK
Each
Estimated Shipment: 04-04-2025
to see stock.
Add to basket
15814764
View Documents
GPE Scientific™
GBM/18/9
Male Spherical Joint 18/9
180.00 DKK
Each
Estimated Shipment: 04-04-2025
to see stock.
Add to basket
15824764
View Documents
GPE Scientific™
GBM/28/12
Male Spherical Joint 28/12
231.00 DKK
Each
Estimated Shipment: 04-04-2025
to see stock.
Add to basket
15834764
View Documents
GPE Scientific™
GBM/28/15
Male Spherical Joint 28/15
253.00 DKK
Each
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
15844764
View Documents
GPE Scientific™
GBM/35/20
Male Spherical Joint 35/20
294.00 DKK
Each
Estimated Shipment: 19-03-2025
to see stock.
Add to basket
15854764
View Documents
GPE Scientific™
GBM/40/25
Male Spherical Joint 40/25
383.00 DKK
Each
Estimated Shipment: 04-04-2025
to see stock.
Add to basket
15864764
View Documents
GPE Scientific™
GBM/51/35
Male Spherical Joint 51/35
489.00 DKK
Each
Estimated Shipment: 04-04-2025
to see stock.
Add to basket
Description
Description
  • Spherical joint designed for use with cup joint
  • Available with different joint sizes, shank bores, and shank ODs
Specifications
Specifications
Male Spherical Joint