Learn More
GPE Scientific™ J Young Spherical Joint, Male/O-Ring Ball Joint
GPE Scientific™ J Young Spherical Joint, Male/O-Ring Ball Joint is an interchangeable spherical joint available with Viton or PTFE-covered Viton O-Ring.
167.00 DKK - 489.00 DKK
Product Code | Brand | Type | Joint Size | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Type | Joint Size | Price | Quantity & Availability | ||||
15884754
|
GPE Scientific™
GBM/12/2 |
Male Spherical Joint | 12/2 |
252.00 DKK
Each |
Estimated Shipment: 04-04-2025
Log in to see stock. |
||||
15894754
|
GPE Scientific™
GBM/12/5 |
Male Spherical Joint | 12/5 |
167.00 DKK
Each |
Estimated Shipment: 04-04-2025
Log in to see stock. |
||||
15804764
|
GPE Scientific™
GBM/18/7 |
Male Spherical Joint | 18/7 |
180.00 DKK
Each |
Estimated Shipment: 04-04-2025
Log in to see stock. |
||||
15814764
|
GPE Scientific™
GBM/18/9 |
Male Spherical Joint | 18/9 |
180.00 DKK
Each |
Estimated Shipment: 04-04-2025
Log in to see stock. |
||||
15824764
|
GPE Scientific™
GBM/28/12 |
Male Spherical Joint | 28/12 |
231.00 DKK
Each |
Estimated Shipment: 04-04-2025
Log in to see stock. |
||||
15834764
|
GPE Scientific™
GBM/28/15 |
Male Spherical Joint | 28/15 |
253.00 DKK
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
||||
15844764
|
GPE Scientific™
GBM/35/20 |
Male Spherical Joint | 35/20 |
294.00 DKK
Each |
Estimated Shipment: 19-03-2025
Log in to see stock. |
||||
15854764
|
GPE Scientific™
GBM/40/25 |
Male Spherical Joint | 40/25 |
383.00 DKK
Each |
Estimated Shipment: 04-04-2025
Log in to see stock. |
||||
15864764
|
GPE Scientific™
GBM/51/35 |
Male Spherical Joint | 51/35 |
489.00 DKK
Each |
Estimated Shipment: 04-04-2025
Log in to see stock. |
||||
Description
- Spherical joint designed for use with cup joint
- Available with different joint sizes, shank bores, and shank ODs
Specifications
Male Spherical Joint |