Learn More
GPE Scientific™ J Young Spherical Joint, Male/O-Ring Ball Joint
GPE Scientific™ J Young Spherical Joint, Male/O-Ring Ball Joint is an interchangeable spherical joint available with Viton or PTFE-covered Viton O-Ring.
Brand: GPE Scientific™ GBM/35/20
This item is not returnable.
View return policy
Description
- Spherical joint designed for use with cup joint
- Available with different joint sizes, shank bores, and shank ODs
Specifications
Shank Bore: 20 mm, Shank OD: 24 mm | |
Male Spherical Joint | |
35/20 |