Learn More
GPE Scientific™ J Young Spherical Joint, Male/O-Ring Ball Joint
GPE Scientific™ J Young Spherical Joint, Male/O-Ring Ball Joint is an interchangeable spherical joint available with Viton or PTFE-covered Viton O-Ring.
Brand: GPE Scientific™ GBM/51/35
This item is not returnable.
View return policy
Description
- Spherical joint designed for use with cup joint
- Available with different joint sizes, shank bores, and shank ODs
Specifications
Shank Bore: 35 mm, Shank OD: 38 mm | |
Male Spherical Joint | |
51/35 |