Learn More
GPE Scientific™ J Young Spherical Joint, Male/O-Ring Ball Joint
GPE Scientific™ J Young Spherical Joint, Male/O-Ring Ball Joint is an interchangeable spherical joint available with Viton or PTFE-covered Viton O-Ring.
Brand:  GPE Scientific™ GBM/12/5
 View more versions of this product
Product Code. 15894754
  • 167.00 DKK / Each
Estimated Shipment: 07-04-2025
to see stock.
Explore more special offers
Description
  • Spherical joint designed for use with cup joint
  • Available with different joint sizes, shank bores, and shank ODs
Specifications
Specifications
Shank Bore: 5 mm, Shank OD: 8 mm
Male Spherical Joint
12/5