Learn More
LAMR1 RPSA Mouse anti-Human, Unconjugated, Clone: 2A9A1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67324-1-IG-20UL
 View more versions of this product
Product Code. 16809895
  • 1081.84 DKK / 20µL
This item is not returnable. View return policy
Description
Description
The LAMR1, RPSA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LAMR1, RPSA. This antibody recognizes human, mouse, rat antigen. The LAMR1, RPSA antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
LAMR1 RPSA
Monoclonal
1000 ÎĽg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot
2A9A1
Unconjugated
20 ÎĽL
3921
IgM
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
LAMR1 RPSA Mouse anti-Human, Unconjugated, Clone: 2A9A1, Proteintech > 20 ÎĽL