missing translation for 'onlineSavingsMsg'
Learn More

LAMR1 RPSA Mouse anti-Human, Unconjugated, Clone: 2A9A1, Proteintech

Mouse Monoclonal Antibody

1081.84 DKK - 3058.99 DKK

Specifications

Antigen LAMR1 RPSA
Clone 2A9A1
Concentration 1000 ÎĽg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16809895
View Documents
Proteintech
67324-1-IG-20UL
20 ÎĽL
1081.84 DKK
20µL
Please to purchase this item. Need a web account? Register with us today!
16899885
View Documents
Proteintech
67324-1-IG-150UL
150 ÎĽL
3058.99 DKK
150µL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The LAMR1, RPSA antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human LAMR1, RPSA. This antibody recognizes human, mouse, rat antigen. The LAMR1, RPSA antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

LAMR1 RPSA
1000 ÎĽg/mL
Monoclonal
Mouse
3921
Primary
2A9A1
Western Blot
Unconjugated
Human, Rat, Mouse
IgM
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
LAMR1 RPSA Mouse anti-Human, Unconjugated, Clone: 2A9A1, Proteintech

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.