Learn More
Lansoprazole, MedChemExpress
MedChemExpress Lansoprazole (AG 1749) is an orally active proton pump inhibitor which prevents the stomach from producing acid. Lansoprazole (AG 1749) is a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor).
Brand: Medchem Express HY-13662

Description
Lansoprazole (AG 1749) is an orally active proton pump inhibitor which prevents the stomach from producing acid. Lansoprazole (AG 1749) is a potent brain penetrant neutral sphingomyelinase (N-SMase) inhibitor (exosome inhibitor).Specifications
Lansoprazole | |
White | |
C16H14F3N3O2S | |
AG-1749 | |
O=S(C1=NC2=CC=CC=C2N1)CC3=NC=CC(OCC(F)(F)F)=C3C | |
369.36 | |
Research | |
Metabolism-protein/nucleotide metabolism |
103577-45-3 | |
Research | |
5 g | |
DMSO : 100 mg/mL (270.74 mM; Need ultrasonic) ∣H2O : 0.1 mg/mL (0.27 mM; Need ultrasonic) | |
369.36 | |
98.0% | |
Solid |
Product Suggestions
Customers who viewed this item also viewed
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Research purposes only