Learn More
Gilac™ Large Volume Container with Pivoting Wheels and Brakes
Large Volume Container with Pivoting Wheels and Brakes is the economical solution for moving large volume containers around.
Brand: Gilac™ G147121
This item is not returnable.
View return policy
Description
- The economical solution for moving large volume containers around.
- Stainless steel wheels and fastenings.
- Stainless steel recommended in humid environments (no damage to bearings even in the presence of water).
- Load capacity for information purposes only (depending on how the load is spread and the operating temperature).
- Bottom with gaskets on the axle for perfect sealing.
- Volume in litres is equivalent to the container filled to the brim.
Specifications
Storage Tote | |
White | |
44.9 gal. | |
4 Pivoting Wheels, 2 Brakes |
HDPE | |
Rectangular | |
170 L |