missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Large Volume Container with Pivoting Wheels and Brakes

Large Volume Container with Pivoting Wheels and Brakes is the economical solution for moving large volume containers around.

5485.00 DKK - 9790.00 DKK

Specifications

Type Storage Tote
Material HDPE
Color White
Shape Rectangular
View More Specs

Products 10
Product Code Brand Capacity (Metric) Includes Price Quantity & Availability  
Product Code Brand Capacity (Metric) Includes Price Quantity & Availability  
17978811
View Documents
Gilac™
G147031
100 L 2 Pivoting Wheels with Brakes
5485.00 DKK
Each
Please to purchase this item. Need a web account? Register with us today!
17948621
View Documents
Gilac™
G147021
100 L 4 Pivoting Wheels, 2 Brakes
5485.00 DKK
Each
Please to purchase this item. Need a web account? Register with us today!
17908821
View Documents
Gilac™
G147131
170 L 2 Pivoting Wheels with Brakes
5875.00 DKK
Each
Please to purchase this item. Need a web account? Register with us today!
17968411
View Documents
Gilac™
G147121
170 L 4 Pivoting Wheels, 2 Brakes
5875.00 DKK
Each
Please to purchase this item. Need a web account? Register with us today!
17998881
View Documents
Gilac™
G147231
220 L 2 Pivoting Wheels with Brakes
6170.00 DKK
Each
Please to purchase this item. Need a web account? Register with us today!
17948961
View Documents
Gilac™
G147221
220 L 4 Pivoting Wheels, 2 Brakes
6170.00 DKK
Each
Please to purchase this item. Need a web account? Register with us today!
17918501
View Documents
Gilac™
G146831
310 L 2 Pivoting Wheels with Brakes
7310.00 DKK
Each
Please to purchase this item. Need a web account? Register with us today!
17928621
View Documents
Gilac™
G146821
310 L 4 Pivoting Wheels, 2 Brakes
7310.00 DKK
Each
Please to purchase this item. Need a web account? Register with us today!
17988881
View Documents
Gilac™
G146931
500 L 2 Pivoting Wheels with Brakes
9790.00 DKK
Each
Please to purchase this item. Need a web account? Register with us today!
17908891
View Documents
Gilac™
G146921
500 L 4 Pivoting Wheels, 2 Brakes
9790.00 DKK
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The economical solution for moving large volume containers around.
  • Stainless steel wheels and fastenings.
  • Stainless steel recommended in humid environments (no damage to bearings even in the presence of water).
  • Load capacity for information purposes only (depending on how the load is spread and the operating temperature).
  • Bottom with gaskets on the axle for perfect sealing.
  • Volume in litres is equivalent to the container filled to the brim.
Specifications

Specifications

Storage Tote
HDPE
White
Rectangular
Videos
SDS
Documents

Documents

Product Certifications