Learn More
Gilac™ Large Volume Container with Pivoting Wheels and Brakes
Large Volume Container with Pivoting Wheels and Brakes is the economical solution for moving large volume containers around.
Brand:  Gilac™ G147231
 View more versions of this product
Product Code. 17998881
  • 6170.00 DKK / Each
Explore more special offers
Description
Description
  • The economical solution for moving large volume containers around.
  • Stainless steel wheels and fastenings.
  • Stainless steel recommended in humid environments (no damage to bearings even in the presence of water).
  • Load capacity for information purposes only (depending on how the load is spread and the operating temperature).
  • Bottom with gaskets on the axle for perfect sealing.
  • Volume in litres is equivalent to the container filled to the brim.
Specifications
Specifications
Storage Tote
White
58.1 gal.
2 Pivoting Wheels with Brakes
HDPE
Rectangular
220 L