Learn More
BRAND™ Lateral Burette Stopcock for Burette, Amber
Lateral Burette Stopcock for Burette includes an interchangeable glass key.
Brand:  BRAND™ 81909
 View more versions of this product
Product Code. 10111971
Description
Description
  • Stopcock housing with precision Borosilicate Glass 3.3 burette tip
  • Interchangeable borosilicate glass key with retention device
  • Stopcock size 3NS/12
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Burette Stopcock
Amber Borosilicate Glass (3.3)
50 mL Burette
Glass Key