Learn More
BRAND™ Lateral Burette Stopcock for Burette, Amber
Lateral Burette Stopcock for Burette includes an interchangeable glass key.
Brand: BRAND™ 81907
This item is not returnable.
View return policy
Description
- Stopcock housing with precision Borosilicate Glass 3.3 burette tip
- Interchangeable borosilicate glass key with retention device
- Stopcock size 3NS/12
Specifications
Burette Stopcock | |
Amber Borosilicate Glass (3.3) | |
25 mL Burette | |
Glass Key |