Learn More
Kautex™ LDPE Narrow Neck Wash Bottle Closures
Top off Series 301 and Series 312 round, narrow neck bottles with Kautex™ LPDE Narrow Neck Wash Bottle Closures. Their color is natural, which matches the bottle color.
Brand:  Kautex™ 2000075993
 View more versions of this product
Product Code. 15961431
This item is not returnable. View return policy
Description
Description
  • LDPE
  • Natural color
  • Round
  • Wash bottle style
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Natural
Wash Bottle Closure
25 mm
LDPE
500 mL Bottle
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kautex™ LDPE Narrow Neck Wash Bottle Closures > Cap Size: 25 mm O.D.; For Use With: 500 mL Bottle