missing translation for 'onlineSavingsMsg'
Learn More
Lithium chloride, Free-flowing, ACS reagent, anhydrous, ≥99%, Honeywell Fluka™ Product Code.: 15687010

Lithium chloride, Free-flowing, ACS reagent, anhydrous, ≥99%, Honeywell Fluka™

Product Code. 15687010 Shop All Honeywell Fluka Products
6 x 500 g, Poly Bottle, Pack of 6
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
6 x 500 g
Packaging:
Poly Bottle
Unit Size:
100g
1kg
2.50kg
500g
Pack of 6
This item is not returnable. View return policy

Product Code. 15687010

Brand: Honeywell-Fluka 7464606X500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Free-flowing, ACS reagent, anhydrous,≥99%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-41-8
Molecular Formula ClLi
Molecular Weight (g/mol) 42.39
MDL Number MFCD00011078
InChI Key KWGKDLIKAYFUFQ-UHFFFAOYSA-M
Synonym lithium chloride, lithium chloride licl, lithiumchloride, licl, chlorure de lithium, chlorku litu, chlorolithium, lithiumchlorid, chlorku litu polish, luthium chlorideShow MoreShow Less
PubChem CID 433294
ChEBI CHEBI:48607
IUPAC Name lithium;chloride
SMILES [Li+].[Cl-]

Specifications

Boiling Point 1,360°C
Melting Point 605°C (literature)
CAS Max % ≤100.0000%
Physical Form Anhydrous
Quantity 6 x 500 g
Linear Formula LiCl
Formula Weight 42.39
Percent Purity >99%
Grade ACS Reagent
Packaging Poly Bottle
Density 2.07 g/cm3
Chemical Name or Material Lithium chloride, Free-flowing, ACS reagent, anhydrous, ≥99%
Show More Show Less
compliance-icons
Product Identifier
  • Lithium chloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lithium chloride, Free-flowing, ACS reagent, anhydrous, ≥99%, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.