Learn More
Heinz Herenz LMS-Swab, Amies
Test tube with the pink screw cap containing 1 mL of Amies medium for sampling and preservation of aerobic, anaerobic, and fastidious bacteria in 48 hours.
Brand: Heinz Herenz 1020203
This item is not returnable.
View return policy
Description
- For transportation and preservation of microbiological samples
- Liquid media allows for easy dilution of samples
- Carry out differnt tests out of one sample
- Ready-to-use device delivered in an easy-to-open peel pouch
- Breakpoint in the shaft if the applicator allows for a safe and hygienic work-flow
- Pre-labeled test tubes allow for correct identification
- Produced according to GMP (Good Manufacturing Practice) and certified according to DIN EN ISO 13485 ensure the high quality
- Sterile (SAL 10-6)
Specifications
Transport and Collection Swab | |
Pink | |
Test tube with screw cap and liquid media | |
Foam stick |
Sterile | |
Pyrogen-free and non-cytotoxic | |
Peel pouch | |
Standard |
Product Content Correction
Product Title
Heinz Herenz LMS-Swab, Amies > Tip: Standard
Spot an opportunity for improvement?Share a Content Correction