Learn More
Lonza™ BioWhittaker™ BME with Earle's BSS, without L-glutamine
Historical precursor to Minimum Essential Medium (MEM)
Brand: Lonza™ BE12-105F
This item is not returnable.
View return policy
Description
- Suitable for variety of cell types
- Manufactured in accordance with cGMP regulations
- Formulations recommended by the Tissue Culture Association
Specifications
15° to 30°C | |
Basal Medium | |
No Glutamine | |
500 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lonza™ BioWhittaker™ BME with Earle's BSS, without L-glutamine > BME with Earle's BSS, without L-glutamine; Quantity: 500mL
Spot an opportunity for improvement?Share a Content Correction