Learn More
Lonza™ FlashGel™ System Sample Preparation Reagents
For use with the Lonza FlashGel System. FlashGel™ System Sample Preparation Reagents for the FlashGel™ System provide the fastest separations and enable the user to watch as nucleic acids migrate in the gel.
Brand:  Lonza™ 50462
 View more versions of this product
Product Code. 11537805
  • 2580.00 DKK / Pack of 5
This item is not returnable. View return policy
Description
Description
  • Reagents include Recovery Buffer and Loading Dye
  • Loading Dye: 5X concentration; RNA native sample buffer
  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals
TRUSTED_SUSTAINABILITY
Specifications
Specifications
5X
Liquid
Room Temperature
DNA Loading Dye
5 x 1 mL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lonza™ FlashGel™ System Sample Preparation Reagents > FlashGel Loading Dye