missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ FlashGel™ System Sample Preparation Reagents

For use with the Lonza FlashGel System. FlashGel™ System Sample Preparation Reagents for the FlashGel™ System provide the fastest separations and enable the user to watch as nucleic acids migrate in the gel.

2580.00 DKK - 2745.00 DKK

Specifications

Form Liquid
Storage Requirements Room Temperature
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11567865
View Documents
Lonza™
57060
2 x 500 μL
2745.00 DKK
Pack of 2
Please to purchase this item. Need a web account? Register with us today!
11537805
View Documents
Lonza™
50462
5 x 1 mL
2580.00 DKK
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Reagents include Recovery Buffer and Loading Dye
  • Loading Dye: 5X concentration; RNA native sample buffer

  • For research or manufacturing use only
  • Not for use in in vitro diagnostic procedures, drug use, or for administration to humans or animals

Specifications

Specifications

Liquid
Room Temperature
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 12 Months

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Lonza™ FlashGel™ System Sample Preparation Reagents

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.