missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ SimplyLoad™ DNA Ladders

Ready-to-load markers contain loading buffer to save time and pipetting steps

Brand:  Lonza™ 50326

 View more versions of this product

Product Code. 11500787

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

•    Consist of RNA transcripts in sizes of 0.5, 1, 1.5, 2, 2.5, 3, 4, 5, 6, and 9kb
•    Can be denatured with standard procedures
•    Can be visualized on Northern blots with labeled lambda sequence
•    Can be detected with ethidium bromide or nucleic acid stains
•    Store at -20°C for up to six months or at -80°C for up to 24 months

TRUSTED_SUSTAINABILITY
Specifications

Specifications

60 ng/μL
Liquid
Gel electrophoresis
50
2° to 8°C
DNA Molecular Weight Marker/Ladder
50% GC content
20bp
500 μL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers