missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ SimplyLoad™ DNA Ladders
Ready-to-load markers contain loading buffer to save time and pipetting steps
2546.01 DKK - 2690.00 DKK
Specifications
Product Type | DNA Molecular Weight Marker/Ladder |
---|---|
Form | Liquid |
Formulation | 50% GC content |
For Use With (Application) | Gel electrophoresis |
Quantity | 500 μL |
Product Code | Brand | Concentration | No. of Bands | Increments | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Concentration | No. of Bands | Increments | Price | Quantity & Availability | |||
11500787
|
Lonza™
50326 |
60 ng/μL | 50 | 20bp | N/A | ||||
11530787
|
Lonza™
50329 |
60 ng/μL | 16 | 500bp | N/A | ||||
11520787
|
Lonza™
50328 |
60 ng/μL | 30 | 20bp |
2690.00 DKK
Each |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||
11510787
|
Lonza™
50327 |
32 ng/μL | 10 | 100bp |
2690.00 DKK
Each |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||
11570787
|
Lonza™
50333 |
60 ng/μL | 21 | 100bp and 1000bp |
2690.00 DKK
Each |
Estimated Shipment: 26-11-2024
Log in to see stock. |
|||
11550787
|
Lonza™
50331 |
40 ng/μL | 25 | 20bp | N/A | ||||
Description
- Consist of RNA transcripts in sizes of 0.5, 1, 1.5, 2, 2.5, 3, 4, 5, 6, and 9kb
- Can be denatured with standard procedures
- Can be visualized on Northern blots with labeled lambda sequence
- Can be detected with ethidium bromide or nucleic acid stains
- Store at -20°C for up to six months or at -80°C for up to 24 months
Specifications
DNA Molecular Weight Marker/Ladder | |
50% GC content | |
500 μL |
Liquid | |
Gel electrophoresis | |
2° to 8°C |
Safety and Handling
missing translation for 'shelfLife' : 6 Months