missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ SimplyLoad™ DNA Ladders
Ready-to-load markers contain loading buffer to save time and pipetting steps
Brand: Lonza™ 50331
This item is not returnable.
View return policy
Description
• Consist of RNA transcripts in sizes of 0.5, 1, 1.5, 2, 2.5, 3, 4, 5, 6, and 9kb
• Can be denatured with standard procedures
• Can be visualized on Northern blots with labeled lambda sequence
• Can be detected with ethidium bromide or nucleic acid stains
• Store at -20°C for up to six months or at -80°C for up to 24 months
Specifications
Blue, Cyan | |
DNA Molecular Weight Marker/Ladder | |
50% GC content | |
20bp | |
500 μL |
40 ng/μL | |
Liquid | |
Gel electrophoresis | |
25 | |
2° to 8°C |