Learn More
Bellingham Stanley™ Low Volume Tube
Brand: Bellingham Stanley™ 35-73
This item is not returnable.
View return policy
Description
Specifications
Low Volume Tube | |
Stainless Steel | |
5 x 10 mm |
Product Content Correction
Product Title
Bellingham Stanley™ Low Volume Tube > Size: 5 x 10mm
Spot an opportunity for improvement?Share a Content Correction