Learn More
Bellingham Stanley™ Low Volume Tube
7420.00 DKK
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
11950048
|
Bellingham Stanley™
35-73 |
5 x 10 mm |
7420.00 DKK
Each |
Estimated Shipment: 05-11-2025
Log in to see stock. |
||||||
11974249
|
Bellingham Stanley™
35-72 |
5 x 25 mm |
7420.00 DKK
Each |
Estimated Shipment: 05-11-2025
Log in to see stock. |
||||||
11964249
|
Bellingham Stanley™
35-71 |
5 x 50 mm |
7420.00 DKK
Each |
Estimated Shipment: 05-11-2025
Log in to see stock. |
||||||
11960048
|
Bellingham Stanley™
35-74 |
5 x 5 mm | ||||||||
Description
Specifications
Low Volume Tube | |
Stainless Steel |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Bellingham Stanley™ Low Volume Tube