missing translation for 'onlineSavingsMsg'
Learn More

LW6, MedChemExpress

MedChemExpress LW6 (HIF-1α inhibitor) is a novel HIF-1 inhibitor with an IC50 of 4.4 μM. LW6 decreases HIF-1α protein expression without affecting HIF-1β expression.

825.84 DKK - 5931.95 DKK

Specifications

Chemical Name or Material LW6
CAS 934593-90-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C26H29NO5
View More Specs

Products 5
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16495798
Encompass_Preferred
View Documents
Medchem Express
HY-13671
10 mg
1238.76 DKK
10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16473376
Encompass_Preferred
View Documents
Medchem Express
HY-13671
25 mg
2678.95 DKK
25mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16421957
Encompass_Preferred
View Documents
Medchem Express
HY-13671
50 mg
3958.00 DKK
50mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16408477
Encompass_Preferred
View Documents
Medchem Express
HY-13671
5 mg
825.84 DKK
5mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16406276
Encompass_Preferred
View Documents
Medchem Express
HY-13671
100 mg
5931.95 DKK
100mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LW6 (HIF-1α inhibitor) is a novel HIF-1 inhibitor with an IC50 of 4.4 μM. LW6 decreases HIF-1α protein expression without affecting HIF-1β expression.
Specifications

Specifications

LW6
Off-White
C26H29NO5
DMSO : 25 mg/mL (57.40 mM; Need ultrasonic) ∣DMF : 17.24 mg/mL (39.59 mM; Need ultrasonic)
435.51
98.0%
Solid
934593-90-5
Research
HIF-1α inhibitor LW8
O=C(OC)C1=CC=C(O)C(NC(COC2=CC=C(C3(C4)CC5CC4CC(C5)C3)C=C2)=O)=C1
435.51
Research
Cancer-programmed cell death
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month

SDS
Documents

Documents

Product Certifications

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.