Learn More
Kimble Chase™ Lyophilization Glaesers
Brand:  Kimble Chase™ ZGEFG139001
667.85 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Product Code. 10421045
  • 703.00 DKK / Pack of 100
Estimated Shipment: 05-06-2025
Explore more special offers
Specifications
2 mL
Documents
One moment while we fetch your results.
Product Title
Kimble Chase™ Lyophilization Glaesers > Capacity: 2mL