Learn More
Applied Biosystems™ MagMAX™ Plant RNA Isolation Kit
The MagMAX Plant RNA Isolation Kit is designed for automated high-throughput or manual purification of total RNA from wide variety of plant species and tissue types.
Brand: Applied Biosystems™ A47157
12179.00 DKK valid until 2025-12-31
Use promo code "24359" to get your promotional price.
Description
The MagMAX Plant RNA Isolation Kit is designed for automated high-throughput or manual purification of total RNA from wide variety of plant species and tissue types. It utilizes paramagnetic bead technology, enabling high yields and robust performance. The high binding capacity, uniform particle size, and rapid magnetic response of MagMAX magnetic beads makes the technology ideal for high-throughput automatic nucleic acid purification, as well as for manual nucleic acid purification by low- throughput users. The resulting high quality purified RNA is free of proteins, nucleases, and other contaminants or inhibitors, and can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, and other enzymatic reactions.
• Versatile—suitable for any plant type and part, including woody, polyphenol-rich, and lignified samples
• High yields of pure total RNA
• High quality—isolates intact total RNA, free of inhibitors and gDNA contamination. Isolated RNA is suitable for long term storage at -20°C.
• Flexible—easily adjustable for high-throughput automation platforms and manual processing
Applications
The MagMAX Plant RNA Isolation Kit is designed for fast extraction of high purity RNA suitable for all conventional molecular biology applications, including:
• RT-PCR
• RT-qPCR
• Northern blotting
• Nuclease protection assay
Benefits of MagMAX magnetic bead–based purification
The magnetic bead–based purification format of the kit allows for easy scaling of the number of samples processed either manually with a magnetic stand or with a magnetic particle processor such as the KingFisher Duo Prime or KingFisher Flex processors.
Magnetic beads offer many benefits compared to other technologies for isolating nucleic acids. Beads bind the RNA and DNA more efficiently than glass fiber filters, resulting in higher and more consistent yields. Additionally, because filters and vacuum manifolds are not used, there is no risk of undigested tissue particulates clogging these items during the extraction process. This clogging issue is of particular concern with fibrous plant tissue samples that that are not fully lysed during the disruption and homogenization steps.
Optimized bead surface chemistry
The Dynabeads MyOne Carboxylic Acid utilized in the MagMAX Plant RNA Isolation Kit has an optimized surface chemistry with high specific surface area that allows efficient kinetics and high sensitivity in nucleic acid capture. These aspects make Dynabeads magnetic technology ideal for the recovery of the RNA targets found in a variety of plant samples. Once captured, the RNA is then eluted in 100 μL and ready for downstream applications.
Specifications
• 750 mL Lysis Buffer; room temperature • 30 mL RNA Binding Beads; 4°C • 3.4 mL DNase I; –20°C • 10.5 mL DNase I Reconstitution Buffer; –20°C • 32 mL Manganese Chloride Solution; –20°C • 125 mL DNase I Buffer (2X); –20°C • 1146 mL Wash Solution 1 (concentrated); room temperature • 521 mL Wash Solution 2 (concentrated); room temperature • 210 mL Rebinding Buffer; room temperature • 313 mL Nuclease-free Water; room temperature |
|
Less than 1 hr. when using automated protocol | |
Plant Samples | |
High-throughput Compatible | |
RT-PCR, qPCR, cDNA library construction, NGS, microarray analysis, blot hybridization, Northern blotting, in vitro translation, nuclease protection assays, nucleic acid labeling | |
Room Temperature |
Magnetic Bead | |
100 μL | |
Total RNA | |
KingFisher™ Duo Prime, KingFisher™ Flex, KingFisher™ Apex | |
1000 Preps | |
≤47 μg |