Learn More
Applied Biosystems™ Megaplex™ Primer Pools, Rodent Pools B v3.0
Megaplex™ Primer Pools, Rodent Pools B v3.0
Brand:  Applied Biosystems™ 4444752
Product Code. 12057789
  • 8680.00 DKK / 50 reactions
Explore more special offers
Description
Description
Megaplex™ Primer Pools, Rodent Pool B v3.0 is available under a single part number containing both the Megaplex™ RT Primers and Megaplex™ PreAmp Primers, Rodent Pools B v3.0. Megaplex™ RT Primers, Rodent Pool B contains RT primers for 306 and 135 unique microRNAs for mouse and rat respectively plus 4 species specific controls. For use when assay sensitivity is of the utmost importance, Megaplex™ PreAmp Primers significantly enhance the ability to detect low expressed microRNAs enabling the generation of a comprehensive expression profile using far less sample, as low as 1 ng of input total RNA. The microRNA targets represented in this pool tend be less well characterized, more narrowly expressed and⁄or expressed at a lower level compared to Rodent Pool A. Rodent Pool B is intended for use with the TaqMan Rodent MicroRNA B Array v3.0, however, it can also be used to prepare cDNA for real-time analysis using the individual TaqMan™ MicroRNA Assays.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Store at -20°C.
qPCR
Megaplex™, TaqMan™
50 reactions
50 Reactions
50 Reactions
Low
RT Primers, PreAmp Primers
Primer Pool
Rat, Mouse
miRNA analysis