missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse FGF-9 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
610.00 DKK - 29767.00 DKK
Specifications
Accession Number | P54130 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Gene ID (Entrez) | 14180 |
Molecular Weight (g/mol) | 23.3 |
Name | Mouse FGF-9 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17864083
|
Gibco™
450-30-100UG |
100 μg |
5860.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17874023
|
Gibco™
450-30-10UG |
10 μg |
1478.00 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
30089705
|
Gibco™
450-30-01M |
2 x 500 μg |
29430.00 DKK
Pack of 2 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17874403
|
Gibco™
450-30-1MG |
2 x 500 μg | N/A | ||||||
17894323
|
Gibco™
450-30-250UG |
250 μg |
10320.00 DKK
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17894473
|
Gibco™
450-30-2UG |
2 μg |
610.00 DKK
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17874143
|
Gibco™
450-30-500UG |
500 μg |
17670.00 DKK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17824333
|
Gibco™
450-30-50UG |
50 μg |
3515.00 DKK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein was isolated as a secreted factor that exhibits a growth-stimulating effect on cultured glial cells. In nervous system, this protein is produced mainly by neurons and may be important for glial cell development. Expression of the mouse homolog of this gene was found to be dependent on Sonic hedgehog (Shh) signaling. Mice lacking the homolog gene displayed a male-to-female sex reversal phenotype, which suggested a role in testicular embryogenesis.Specifications
P54130 | |
14180 | |
Mouse FGF-9 | |
-20°C | |
<1 EU/μg | |
FGF9 | |
Unconjugated | |
PLGEVGSYFG VQDAVPFGNV PVLPVDSPVL LNDHLGQSEA GGLPRGPAVT DLDHLKGILR RRQLYCRTGF HLEIFPNGTI QGTRKDHSRF GILEFISIAV GLVSIRGVDS GLYLGMNEKG ELYGSEKLTQ ECVFREQFEE NWYNTYSSNL YKHVDTGRRY YVALNKDGTP REGTRTKRHQ KFTHFLPRPV DPDKVPELYK DILSQS | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
ELISA, Functional Assay, Western Blot | |
23.3 | |
E. coli | |
RUO | |
Eks; elbow knee synostosis; FGF; Fgf9; FGF-9; Fibroblast growth factor; Fibroblast growth factor 9; fibroblast growth factor 9 (glia-activating factor); GAF; glia activating factor; glia-activating factor; HBFG-9; HBGF-9; Heparin-binding growth factor 9; M-FGF-9; SYNS3 | |
Mouse | |
Recombinant | |
Lyophilized |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title