Learn More
BRAND™ Narrow Neck Glass Erlenmeyer Flasks
These Erlenmeyer Flasks are manufactured from Boro 3.3 glass and are acc. to DIN ISO 1773.
Brand: BRAND™ 92717
749.82 DKK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
- With beaded rim and graduation
- Narrow neck for controlled pouring and dispensing
- Differing package counts according to capacity
Specifications
DIN ISO 1773 | |
50 mL | |
90 mm | |
10 | |
Yes |
22 mm | |
Borosilicate Glass (3.3) | |
Narrow | |
Erlenmeyer flask | |
51 mm |
Special Offers
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now