Learn More
Kartell™ Neoprene O-rings
Complete a filtration and vacuum desiccator with a spare o-ring. Kartell™ Neoprene O-rings form a seal with the transparent cover and polypropylene base of a filtration and vacuum desiccator.
Brand: Kartell™ 0055600
This item is not returnable.
View return policy
Description
- Essectial component of a Filtration and Vacuum Desiccator
- Spare O-ring
- Spread a little grease on O-ring for assistance in establishing initial vacuum
Specifications
For 230 | |
Neoprene | |
O-Ring |
Black | |
1Each |
Product Content Correction
Product Title
Kartell™ Neoprene O-rings > ForUseWithEquipment: For 230
Spot an opportunity for improvement?Share a Content Correction