Learn More
Kartell™ Neoprene O-rings
Complete a filtration and vacuum desiccator with a spare o-ring. Kartell™ Neoprene O-rings form a seal with the transparent cover and polypropylene base of a filtration and vacuum desiccator.
28.85 DKK - 55.20 DKK
Specifications
Color Black
Product Type O-Ring
Material Neoprene
Quantity 1Each
View More Specs
Products 3
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
11969898
View Documents
Kartell™
0055700
For 8890A
55.20 DKK
Each
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
12523357
View Documents
Kartell™
0055500
For 8890X
28.85 DKK
Each
Estimated Shipment: 28-05-2025
to see stock.
Add to basket
12034707
View Documents
Kartell™
0055600
For 230
38.95 DKK
Each
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
Description
Description
  • Essectial component of a Filtration and Vacuum Desiccator
  • Spare O-ring
  • Spread a little grease on O-ring for assistance in establishing initial vacuum
Specifications
Specifications
Black
O-Ring
Neoprene
1Each
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Kartell™ Neoprene O-rings