missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nepafenac-d5, MedChemExpress
MedChemExpress Nepafenac D5 (AHR-9434 D5) is the deuterium labeled Nepafenac, which is a selective COX-2 inhibitor.
7921.38 DKK - 12873.51 DKK
Specifications
Chemical Name or Material | Nepafenac-d5 |
---|---|
CAS | 1246814-53-8 |
Color | Yellow |
Purity Grade Notes | Research |
Molecular Formula | C15H9D5N2O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16497298
|
Medchem Express
HY-17357S |
10 mg |
12873.51 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16469807
|
Medchem Express
HY-17357S |
5 mg |
7921.38 DKK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Nepafenac D5 (AHR-9434 D5) is the deuterium labeled Nepafenac, which is a selective COX-2 inhibitor.Specifications
Nepafenac-d5 | |
Yellow | |
C15H9D5N2O2 | |
DMSO : ≥ 100 mg/mL (385.64 mM) | |
259.31 | |
98.0% | |
Solid |
1246814-53-8 | |
Research | |
AHR-9434-d5 AL-6515-d5 | |
O=C(C1=C([2H])C([2H])=C([2H])C([2H])=C1[2H])C2=CC=CC(CC(N)=O)=C2N | |
259.31 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only