missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Usbeck Laborgeraete™ Nickel Plated Steel Forceps
22.00 DKK - 80.25 DKK
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | ||||||
10300802
|
Usbeck Laborgeraete™
3130 |
105 mm |
25.21 DKK
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||||
10750313
|
Usbeck Laborgeraete™
3002 |
130 mm |
22.00 DKK
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||||
10505682
|
Usbeck Laborgeraete™
3005 |
200 mm |
43.11 DKK
Each |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
10639531
|
Usbeck Laborgeraete™
3007 |
300 mm |
80.25 DKK
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||||
Specifications
1 Pack | |
Nickel-plated Steel | |
Forceps |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Usbeck Laborgeraete™ Nickel Plated Steel Forceps