Learn More
Masterflex™ PharMed™ BPT Opaque 2-stopped Pump Tubing
Protect your light-sensitive samples
Brand: Masterflex™ 96114-15
This item is not returnable.
View return policy
Description
- Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
Specifications
4.8 mm | |
-50°C | |
15 | |
3/16in. | |
Opaque | |
132°C |
PharMed BPT | |
4 Pack | |
1.7 to 1000mL/min. | |
8.8 m | |
40 psi | |
2-stop Tubing |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Masterflex™ PharMed™ BPT Opaque 2-stopped Pump Tubing > Tubing Size: 15
Spot an opportunity for improvement?Share a Content Correction