Learn More
Thermo Scientific™ Oxoid™ AnaeroGen™ Compact Sachet
This compact sachet is a simple anaerobic gas generator system for the anaerobic incubation of up to four petri dishes or an identification panel.
Brand:  Thermo Scientific™ AN0020D
345.37 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15219057
  • 559.00 DKK / Pack of 10
Explore more special offers
Description
Description
Thermo Scientific™ Oxoid™ AnaeroGen™ Compact Sachet is a simple anaerobic gas generator system for the anaerobic incubation of up to four petri dishes or an identification panel.
  • Use one sachet / Compact Plastic Pouch
  • Use with Compact Plastic Pouch, Part No. AG0020C and AnaeroGen Compact Sealing Clip, Part No. AN0005C
Includes: 10 sachets.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Compact Sachet
10 Sachets and 10 Pouches
10/Pk.
Compact Gas Generation Sachet
Thermo Scientific Oxoid
SDS
Documents Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number4261MH Certificate TypeCertificate of Analysis Date09/04/2025 Catalog NumberAN0020D
Lot Number4243MH Certificate TypeCertificate of Analysis Date19/02/2025 Catalog NumberAN0020D
Lot Number4221MH Certificate TypeCertificate of Analysis Date03/01/2025 Catalog NumberAN0020D
Lot Number4030MH Certificate TypeCertificate of Analysis Date25/11/2024 Catalog NumberAN0020D
Lot Number4136MH Certificate TypeCertificate of Analysis Date19/11/2024 Catalog NumberAN0020D
Product Title
Thermo Scientific™ Oxoid™ AnaeroGen™ Compact Sachet >