Learn More
p19 INK4d Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10272-2-AP-20UL
 View more versions of this product
Product Code. 16836553
  • 1081.84 DKK / 20µL
This item is not returnable. View return policy
Description
Description
p19 / INK 4d is a specific inhibitor of cdk 4 / cdk 6 and is periodically expressed during cell cycle. It is maximally induced as cells enter S-phase.
Specifications
Specifications
p19 INK4d
Polyclonal
Unconjugated
CDKN2D
CDKN2D, INK4D, P19, p19 INK4D, p19 INK4d
Rabbit
Antigen Affinity Chromatography
RUO
1032, 12581, 494444
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P55273, Q60773
Cdkn2d
p19 INK4d Fusion Protein Ag0415
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
p19 INK4d Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech > 20 μL; Unconjugated