missing translation for 'onlineSavingsMsg'
Learn More
Learn More
p19 INK4d Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1081.84 DKK - 3058.99 DKK
Specifica
Antigen | p19 INK4d |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Codice del prodotto | Marca | Quantity | Prezzo | QuantitĂ e disponibilitĂ | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantity | Prezzo | QuantitĂ e disponibilitĂ | |||||
16836553
|
Proteintech
10272-2-AP-20UL |
20 ÎĽL |
1081.84 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16826553
|
Proteintech
10272-2-AP-150UL |
150 ÎĽL |
3058.99 DKK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Descrizione
p19 / INK 4d is a specific inhibitor of cdk 4 / cdk 6 and is periodically expressed during cell cycle. It is maximally induced as cells enter S-phase.Specifica
p19 INK4d | |
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
P55273, Q60773 | |
1032, 12581, 494444 | |
p19 INK4d Fusion Protein Ag0415 | |
Primary | |
-20°C | |
CDKN2D |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CDKN2D, INK4D, P19, p19 INK4D, p19 INK4d | |
Cdkn2d | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Individuate un'opportunitĂ di miglioramento?Condividi una correzione di contenuto
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
p19 INK4d Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech