missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Paricalcitol-d6, MedChemExpress
MedChemExpress Paricalcitol-d6 is the deuterium labeled Paricalcitol. Paricalcitol is a drug used for the prevention and treatment of secondary hyperparathyroidism (excessive secretion of parathyroid hormone) associated with chronic renal failure.
5938.53 DKK - 47538.39 DKK
Specifications
Chemical Name or Material | Paricalcitol-d6 |
---|---|
CAS | 2070009-67-3 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C27H38D6O3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16498208
|
Medchem Express
HY-76585 |
10 mg |
47538.39 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16481969
|
Medchem Express
HY-76585 |
1 mg |
5938.53 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16470648
|
Medchem Express
HY-76585 |
5 mg |
28525.04 DKK
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Paricalcitol-d6 is the deuterium labeled Paricalcitol. Paricalcitol is a drug used for the prevention and treatment of secondary hyperparathyroidism (excessive secretion of parathyroid hormone) associated with chronic renal failure.Specifications
Paricalcitol-d6 | |
White | |
C27H38D6O3 | |
C[C@]([C@@H]([C@@H](/C=C/[C@@H](C(C([2H])([2H])[2H])(C([2H])([2H])[2H])O)C)C)CC1)(CCC/2)[C@@]1(C2=C\C=C3C[C@H](C[C@@H](C/3)O)O)[H] | |
422.67 | |
Research | |
Solid |
2070009-67-3 | |
Research | |
DMSO : 100 mg/mL (236.59 mM; Need ultrasonic) | |
422.67 | |
99.64% | |
Metabolism-sugar/lipid metabolism |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only