Learn More
ratiolab™ Pasteur-Plast Pipets
Render liquid handling simple and safe with these Pasteur-Plast pipets, which are inert to biological fluids and most acids. The low affinity of the surface reduces the loss of cells and valuable proteins through binding.
Brand:  ratiolab™ 2600145
 View more versions of this product
Product Code. 13407477
  • 390.00 DKK / Pack of 500
Estimated Shipment: 05-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Soft, elastic bulb is unbreakable, thus reducing the risk of glass injury
  • Burr-free orifice for neat, clean and precise dispensing
  • Only externally graduated Pasteur-plast pipets with clearly visible embossed graduations
Specifications
Specifications
LDPE
No
≤70°C
0.2 mL
Pasteur Pipet
Non-sterile
500 Pipets/Box
Clear
130 mm
Single-use
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
ratiolab™ Pasteur-Plast Pipets > Pasteur-Plast Pipets; Graduated: No; Length (Metric): 130 mm; Sterility: Non-sterile; Volume (Metric): 0.2 mL