Learn More
Cole-Parmer™ Phenolic Base Trolley
Safe and effective way to move large carboys, drums, and crates
Brand:  Cole-Parmer™ 09378-05
Product Code. 12375709
  • 8625.00 DKK / Each
Estimated Shipment: 25-07-2025
Explore more special offers
Description
Helps eliminate back strain and heavy lifting; just tip cart forward and slide carboy on the cart. Square steel tubing supports phenolic base with a melamine high pressure laminate surface. Hard rubber castors swivel 360°For easy navigation. Base is 108mm from the floor. Handle is 978mm high and 483mm wide. Safe and effective way to move large carboys, drums, and crates
Specifications
Specifications
Trolley
Phenolic
Product Title
Cole-Parmer™ Phenolic Base Trolley > Material: Phenolic Base