Learn More
DWK Life Sciences Kimble™ Phenolic Screw Cap with Liner
Secure your media during autoclaving with the Kimble™ Phenolic Screw Cap with Liner. It is made of a special phenolic material (wood filled) and very resistant to the effects of temperature and steam encountered in autoclaving
Brand:  DWK Life Sciences KSSR08425K#
 View more versions of this product
Product Code. 11357584
  • 373.00 DKK / Pack of 100
Estimated Shipment: 09-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Secure your media during autoclaving with the Kimble™ Phenolic Screw Cap with Liner. It is made of a special phenolic material (wood filled) and very resistant to the effects of temperature and steam encountered in autoclaving
  • Styrene Liner
  • Screw Cap
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Phenolic
100 Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
DWK Life Sciences Kimble™ Phenolic Screw Cap with Liner >