Learn More
Memmert™ Plug-In Tube Extension for SF110, Straight
Plug-in tube extension designed for exhaust air ducting and connection by hose if necessary.
Brand:  Memmert™ SF110B29719
Product Code. 17598065
  • 938.00 DKK / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Description
  • Accessory
  • Plug-in tube extension (outer diameter 60.3 mm, inner 57 mm)
  • Straight
Specifications
Specifications
60.3 mm
SF110 Sterilizer
Product Title
Memmert™ Plug-In Tube Extension for SF110, Straight > For Use With: SF110