Learn More
Brady™ Polyester Labels for M610, M710, M611
Polyester Labels for use with Brady™ Printers M611, M610 and M710.
Brand:  Brady™ 312184
 View more versions of this product
Product Code. 17818004
  • 296.00 DKK / Each
In Stock
to see stock.
Explore more special offers
Description
  • Terminal block markers for the M610 and M611 Thermal Transfer Portable Printers
  • This material is UL approved with its respective ribbon
  • Parts can also be printed on the M710 Label Printer provided the roll adapter (M71-ADAPT) is used
Specifications
Specifications
Permanent Acrylic
Transparent
Label
Single-side Printable
Labels
Data and Telecommunications Labeling, Electrical Labeling
1 Label
1 Label
-20°C to +70°C
0.051 mm
49.21 ft.
0.18 in.
4.5 mm
None
Matte
Polyester
Thermal Transfer
Non-sterile
M610, M710, M611, BMP61, BMP71
-20°C
-4°F to +158°F
0.002 in.
15 m x 4.5 mm
15 m
4.5 mm