missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Degree of polymerization ca. 140 - 170
- Hydroxyl value 17.0 - 21.0
- Storage Temperature: +15 to +30°C
Specifications
Specifications
CAS | 25322-68-3 |
Color | White |
pH | 4.5-7.5 |
Quantity | 500 g |
Chemical Name or Material | Glycol 6000 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Polyethylene Glycol 6000, Serva Electrophoresis™
Spot an opportunity for improvement?Share a Content Correction