Learn More
Vitlab™ Polypropylene Spray Bottles
Use these bottles for spraying detergents or disinfectants, especially into difficultly accessible areas, as well as applications in thin layer chromatography. VITLAB™ Polypropylene Spray Bottles feature a sprayer insert with stable, smoothly operated pump trigger and adjustable spray nozzle.
Brand: Vitlab™ 53610
Description
- White polypropylene spray bottles
- Spray nozzle can be regulated from the finest mist (nebulizing) to a precise liquid jet
- Range: approx. 3 to 4 meters
Specifications
850 mL | |
Ideal for spraying detergents or disinfectants, especially into difficultly accessible areas, as well as applications in thin layer chromatography | |
5 Pack | |
Yes | |
Spray Bottle | |
Screw Cap |
White | |
Polypropylene | |
White | |
Not Autoclavable | |
Polypropylene |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For research purposes only