Learn More
Ratiolab™ Polypropylene Urine Beakers
Collect urine specimens in these beakers. Ratiolab™ Polypropylene Urine Beakers are ideal for sampling, transport and storage of solid, pasty and liquid samples.
Brand:  Ratiolab™ 5921182
 View more versions of this product
Product Code. 10369182
  • 549.00 DKK / Pack of 500
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Description
  • Available in polypropylene (PP) or polystyrene (PS)
  • Practical multi-purpose beakers and bottles
  • Ideal for the clinical routine
  • With red screw-cap
  • Supplied in non-sterile or sterile, individually packed
Specifications
Specifications
500 Pk.
125 mL
Urine Beaker
Polypropylene
Documents
One moment while we fetch your results.
Product Title
Ratiolab™ Polypropylene Urine Beakers > Capacity: 125mL