Learn More
WTW™ ProfiLine™ pH 3310 Portable pH Meter
Portable, waterproof and robust meters for the determination of the parameters pH, ORP, dissolved oxygen, conductivity and BSB
Brand:  WTW™ 2AA312
7572.87 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10186573
  • 7765.00 DKK / Each
Explore more special offers
This item is not returnable. View return policy
Description
Description
The pH 3310 is an elegant combination of portable meter and data logger for all those who want to save measuring series automatically and keep processing them on a computer. The meter includes a built-in data memory for 5000 GLP-compliant entries with an interval-controlled logger. All data can conveniently be transmitted to a computer via the USB interface. The CMC function visualizes the optimum measuring range and thus supports correct measurements. The graphic display with clear text menus allows the comfortable and safe operation of the meter.
TRUSTED_SUSTAINABILITY
Specifications
Show More
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ ProfiLine™ pH 3310 Portable pH Meter > Includes: Data logger and USB interface, carry case with pH electrode SenTix 41